SP Bel-Art Tubing Cutter, Plastic, Bel-Art Products, a part of SP
Catalogue # 63022-100
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Tubing Cutter
- Material:Plastic
- Cat. no.:63022-100
- Cutting Capacity:1.9 cm (¾")
- Overall Length:7.6 cm (3")
- Supplier No.:210100000
Spécifications
À propos de cet article
Pocket-sized cutter cuts flexible tubing with an outer diameter up to 19 mm (3/4").
- Hardened steel blade
- Jaw is spring-loaded and closes automatically
- Compact and convenient size
The jaw is lightly spring-loaded and closes automatically for safety. The replaceable, hardened steel blade produces clean, straight cuts.
Ordering information: Replacement blades sold separately.