SP Bel-Art Economy Sampling Scoops, Bel-Art Products, a part of SP
Designed for accurate measurement of small quantities of dry or liquid substances.
- One-piece design
- Polystyrene construction
- Durable
- Lightweight
- Flat-bottom shape
One-piece scoops are molded from durable, FDA-grade, high-impact polystyrene plastic. Leveling the sample to the flat top edge provides accurate and repeatable volume measurements. Flat-bottom shape and light weight allow use as a weighing boat.
Frequently Bought Together