SP Bel-Art Stainless Steel Reagent Digger, Bel-Art Products, a part of SP
Catalog # 62489-110
Supplier: Bel-Art Products, a Part of SP
Specifications
- Length:22.9L cm (9")
- Cat. no.:62489-110
- Supplier No.:367130000
Specifications
About this item
Ideal for dislodging and retrieving stubborn, caked, or crystallized materials from deep down inside a bottle or vessel that has a minimum neck diameter of 24 mm (15/16").
- Ergonomically designed
- Suitable for a variety of reagent applications
- Stainless steel construction
- Ideal for removing stubborn materials from deep vessels
This utensil is ergonomically designed with a curved shape, one pointed end, and one blunt end. The shape facilitates scooping, shoveling, digging, and transferring of powdered, flaked, crystalline, or granular media.