VWR® Beaker Tongs
24.1 cm (9½"), Stainless steel
Supplier: VWR International
Jump to:
Product Details & Documents
Tongs feature a three-prong design and carbon steel, cadmium-plated construction.
- Carbon steel, cadmium-plated construction
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 mL capacities.
Specifications
- Longueur:24.1 cm (9½")
- Matière:Stainless steel