Gamma Irradiation Sterilization Bags, Self-Seal, With Gamma Irradiation Process Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificates
About this item
Gamma irradiation sterilization bags can be used as the primary, secondary, or tertiary packaging for items that will go through gamma irradiation processing or sterilization. Ideal for pharmaceutical and biotech production applications.
- Wide range of sizes
- Convenient self-seal closure; no need for the use of or validation of a heat sealer
- No tools required to open
- Impervious to liquid sanitization agents
- Visibility inside the bag allows for identification of contents
- Prolong the storage and shelf-life of products sterilized 'in-house' or by contract sterilization services
Closure type: Self-adhesive
Latex free: Yes
Opening: Easy tear
Process indicator: Gamma (yellow to red)
Certifications: Certificates of Conformance.
Caution: Gamma irradiation sterilization bags are not autoclavable.
Frequently Bought Together