About this item
This pseudotyped virus uses recombinant vesicular stomatitis virus (rVSV) to carry the S protein of SARS-CoV-2 (GenBank: MN908947) with multiple mutations initially identified in variant of Omicron JN.1. See mutation on our website. The S has 18-aa cytoplasmic tail truncation for optimal infection.
- Applications:
- Anti-SARS-CoV-2 neutralizing antibody screening at high throughput
- Anti-SARS-CoV-2 drug screening at high throughput
- COVID-19 vaccine efficacy evaluation at high throughput
- SARS-CoV-2 pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC analysis
Description: Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See our titration result showing that the starting 2-fold diluted pseudovirus generates signal >50,000-fold higher than uninfected control (cell alone as background in blue). We also evaluated the neutralizing activity of one own S2-specific antibody RV6, with results showing ~ 75% signal reduction.
Biosafety level: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach.
Use protocol: Download complete protocols from 'Product' page at bottom. We strongly suggest that you use small aliquot to test your own systems and use our data as reference. Briefly, incubate 100 μl diluted pseudovirus with 100 μl of your sample in each well of 96-well plate with at least one duplicate for 30 to 60 min at 37 °C. Then add 100 μl target cells (Vero E6 or others expressing ACE2 receptor). Read luciferase signal next day. For protocol using reduced amount, incubate 40 μl diluted virus with 10 μl your sample in each well of 96-well plate for 30 to 60 min at 37 °C. Then add 25 μl target cells. Add 150 μl fresh media next day and read signal on Day 3. To be noted for neutralizing titer (such as EC50) calculation, more duplication leads to more accurate reading.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.