À propos de cet article
Reporter virus particles (RVPs) package the capsid-preMembrane-envelope (C-prM-E) gene construct of West Nile Virus (WNV) (GenBank: DQ211652.1, NY99 strain) and a Renilla luciferase reporter. As the wild typed WNV is handled under BSL-2 precautions, this WNV RVP can be handled using BSL-2 containment practices.
- Applications:
- Anti-West Nile Virus (WNV) neutralizing antibody screening at high throughput
- Anti-West Nile Virus (WNV) drug screening at high throughput
- West Nile Virus (WNV) vaccine efficacy evaluation at high throughput
- West Nile Virus (WNV) pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC or ADE analysis
Description: Infection of cells with this RVP carrying Renilla luciferase reporter results in high level Renilla luciferase activity. See our titration result showing that the starting 2-fold diluted RVP-infected target cells (Vero) generates signal ~ 10,000-fold higher than uninfected control (cell alone as background in blue). We also evaluated the neutralizing activity of one known neutralizing monoclonal antibody- E60 that binds to domain DI/DII region of the viral envelope protein.
Biosafety level: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach.
Use protocol: A complete protocol is supplied with the shipping of product. Briefly, incubate 50 μl diluted virus with your sample in each well of 96-well plate with at least one duplicate for 30 to 60 min at 37 °C. Then add 50 μl target cells (Vero or others expressing receptor). Read Renilla luciferase signal next day.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.