About this item
This pseudotyped virus uses recombinant vesicular stomatitis virus (rVSV) to carry the full length fusion (F) protein of Human metapneumovirus (HMPV) isolate NL/1/99 B1 strain (GenBank: AY525843.1). HMPV is a respiratory virus with fusion (F) protein as the sole target of neutralizing antibodies. As pseudovirus infectivity of rVSV without its original G is restricted to a single round of replication, the pseudotypes can be handled using BSL-2 containment practices.
- Applications:
- Anti-HMPV neutralizing antibody screening at high throughput
- Anti-HMPV drug screening at high throughput
- HMPV vaccine efficacy evaluation at high throughput
- HMPV pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC analysis
Description: Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See our titration result showing that the starting 2-fold diluted pseudovirus generates signal ~ 10,000-fold higher than uninfected control (cell alone as background in blue). An RSV and hMPV cross-reactive mAb RV35 (Cat # M72M26), along with 3 known neutralizing mAbs (101F, D25 and MPE8) were used to validate its neutralizing activity against this pseudovirus.
Biosafety level: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach.
Use protocol: A complete protocol is supplied with the shipping of product. Briefly, incubate 50 μl diluted virus with your sample in each well of 96-well plate for 30 to 60 min at 37 °C. Then add 50 μl target cells, such as Vero E6 (J Clin Microbiol. 45: 1912-1919.), Lung cells or others expressing HMPV receptor, followed by luciferase signal reading.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.