À propos de cet article
This pseudotyped virus uses recombinant vesicular stomatitis virus (rVSV) to carry the full length fusion (F) protein of Human metapneumovirus (HMPV) isolate NL/1/00/A1 (294E). HMPV is a respiratory virus with fusion (F) protein as the sole target of neutralizing antibodies. As pseudovirus infectivity of rVSV without its original G is restricted to a single round of replication, the pseudotypes can be handled using BSL-2 containment practices.
- Applications:
- Anti-HMPV neutralizing antibody screening at high throughput
- Anti-HMPV drug screening at high throughput
- HMPV vaccine efficacy evaluation at high throughput
- HMPV pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC analysis
Description: Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See our titration result showing that the starting 2-fold diluted pseudovirus generates signal ~ 10,000-fold higher than uninfected control (cell alone as background in blue). An RSV and hMPV cross-reactive mAb RV35, along with a known neutralizing mAb (D25) were used to validate its neutralizing activity against this pseudovirus.
Biosafety level: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach.
Use protocol: A complete protocol is supplied with the shipping of product. Briefly, incubate 50 μl diluted virus with your sample in each well of 96-well plate for 30 to 60 min at 37 °C. Then add 50 μl target cells, such as Vero E6 (J Clin Microbiol. 45: 1912-1919.), lung cells or others expressing HMPV receptor, followed by luciferase signal reading.