À propos de cet article
A recombinant trimeric form of the HIV-1 glycoprotein (GP protein) from BG505 strain (clade A, GenBank: ABA61516.1) was produced by human embryonic kidney HEK293 cells, followed by Lectin purification. This product contains ectodomain of GP protein (end at D664) with furin cleavage site replaced by a linker, a T332N mutation for glycan, a I559P mutation for stable trimer formation, and C-terminal 8 hexa-histidine tag.
This product has been evaluated by Size Exclusion Chromatography (SEC) to confirm the trimer formation, SDS-PAGE gel for purity and ELISAs for binding, see results. Due to the unique feature of BG505 at residue 241 glycan hole, a rabbit antibody 11B can target this protein (ref: Cell Rep. 2016;16(9):2327-38.), but not other HIV GP with 241N, such as YU2 or 16055 trimer.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Storage conditions:–80 °C
- Endotoxin-free:No
- Carrier-free:Yes
- Protease-free:No
- Animal-free:Yes
- Protein/peptide name:HIV-1 Trimeric GP140 protein-clade A, BG505 strain
- Molecular weight:~ 98 kDa
- Concentration:0.2 mg/ml
- Formulation:Frozen Liquid. In PBS pH 7.5
- Nuclease-free:No
- Shipping temperature:Dry Ice
- Tested applications:Crystallization (Crys), ELISA, SDS-PAGE (SDS), Western Blotting (WB), Immunization or Antibody Sorting Probe