ContactUsLinkComponent
Stilla® Ruby Chip
Stilla® Ruby Chip
Catalog # 77392-266
CAS Number:  
undefined
Stilla® Ruby Chip
Catalog # 77392-266
Supplier Number:  C16011
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ruby chip
  • Cat. no.:
    77392-266
  • Units:
    Pack of 12 chips
  • Details:
    Storage temperature: −20... ±5 °C
Specifications
About this item
The Ruby Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single fully automation-compatible consumable.
  • Capacity: 16 samples/chip
  • Input volume: 5 µl/chamber
  • Droplets per sample: Up to 17000
  • Dynamic range: ∼4.7 log
  • Fully compatible with integration to automated liquid handling equipment
For higher throughput applications, the Ruby Chip offers the analysis of up to 48 samples per run for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the naica® system or the Nio™+ for partitioning, PCR amplification, and sample read-out. The Ruby Chip is also fully compatible with integration to automated liquid handling equipment.
Digital PCR Consumables and Reagents
Product Information