Epic High Performance Static Dissipative Frocks
À propos de cet article
Lint-free film coated frock for use in critical, ESD and low particulate clean environments. Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas.
- Snap front closures
- Knit wrists
- Knit collar
- No pockets
Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane. The finished fabric contains a proprietary additive making it static dissipative. Individually inspected for sizing, stitching and workmanship.
White garment with white snaps, white knit wrists, and white knit collar.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case.
9 Options Available Below
Product Details & Documents
Lint-free film coated frock for use in critical, ESD and low particulate clean environments. Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas.
- Snap front closures
- Knit wrists
- Knit collar
- No pockets
Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane. The finished fabric contains a proprietary additive making it static dissipative. Individually inspected for sizing, stitching and workmanship.
White garment with white snaps, white knit wrists, and white knit collar.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case.
Specifications for Product Family
Spécifications
- Catalog No:
- 77394-772
- 77394-774
- 77394-776
- 77394-778
- 77394-780
- 77394-782
- 77394-784
- 77394-786
- 77394-788
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionAvailabilityPrice
- Supplier Number: 846855-SSpécifications:
More
Specifications
Cat. No.77394-772Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-MSpécifications:
More
Specifications
Cat. No.77394-774Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-LSpécifications:
More
Specifications
Cat. No.77394-776Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-XLSpécifications:
More
Specifications
Cat. No.77394-778Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-2XLSpécifications:
More
Specifications
Cat. No.77394-780Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-3XLSpécifications:
More
Specifications
Cat. No.77394-782Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-4XLSpécifications:
More
Specifications
Cat. No.77394-784Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-5XLSpécifications:
More
Specifications
Cat. No.77394-786Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong - Supplier Number: 846855-6XLSpécifications:
More
Specifications
Cat. No.77394-788Closure typePress studs / snapsPocketsWithout pocketsSleeve or leg lengthLong