HEPES Buffered Saline Solution, 100 ml
HEPES buffered saline solution
Supplier #: CC-5022
HEPES-Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.
HEPES-BSS is a component of the ReagentPack® Subculture kit (catalog number CC-5034).
Delivery information: Content: 1 × Bottle HEPES Buffered Solution 100 ml.
- Désignation:HEPES buffered saline solution