HEPES Buffered Saline Solution, 100 ml
HEPES Buffered Saline Solution, 100 ml
HEPES buffered saline solution
  CA10064-048
Supplier #:  CC-5022
HEPES Buffered Saline Solution, 100 ml
  CA10064-048
 :  
HEPES Buffered Saline Solution, 100 ml
HEPES Buffered Saline Solution, 100 ml
HEPES buffered saline solution
 :  
$95.21
Each
Minimum order quantity is 1
HEPES-Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.
HEPES-BSS is a component of the ReagentPack® Subculture kit (catalog number CC-5034).
Delivery information: Content: 1 × Bottle HEPES Buffered Solution 100 ml.
  • Désignation:
    HEPES buffered saline solution