NHDC - Human Dendritic Cells | Avantor
NHDC - Human Dendritic Cells
NHDC - Human dendritic cells, 2.5 million cells
Catalog # CA10064-282
Fournisseur #:  CC-2701
NHDC - Human Dendritic Cells
Catalog # CA10064-282
CAS Number:  
NHDC - Human Dendritic Cells
NHDC - Human Dendritic Cells
NHDC - Human dendritic cells, 2.5 million cells
CAS Number:  
Price
$2,018.29
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Dendritic Cells (DCs) act as the messenger cells of the immune system by processing antigens from pathogens and presenting the antigens to T cells to initiate an immune response.
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Poietics® Normal Human Dendritic Cells are tested for a panel of specific surface markers, including CD11c+, CD86+, CD80+. HLADR+, and CD14- to provide detailed characterization of the cells. These cells can survive up to 7 days in culture with the proper cytokines (IL-4 and GM-CSF) to maintain the cells as immature dendritic cells (iDCs). Cells can be further differentiated to mature dendritic cells with the addition of TNF-α and IL-1β.
Applications: Antigen presentation and uptake Primary Research Pharmacology Drug-induced immunosuppression of cytokine secretion.
Cryopreserved Poietics® Normal Human Dendritic Cells (NHDC) are guaranteed to contain ≥3 million viable cells per ampoule. Expression levels are tested for CD80, CD86, HLA-DR, CD11c, and CD14 and provided for information only. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Recommended Medium: LGM-3® Lymphocyte Growth Medium + 50 ng/ml GM-CSF + 50 ng/ml IL-4.
Delivery information: Content: 1× Cryopreserved ampule of NHDC containing ≥2.5 million cells. Certificate of Analysis provided for every cell lot.
Spécifications
Spécifications
  • Désignation:
    NHDC - Human dendritic cells
  • Taille:
    2.5 million cells