PrEC – Human Prostate Epithelial Cells | Avantor
PrEC – Human Prostate Epithelial Cells
PrEC – Human prostate epithelial cells, ≥500000 cells
Catalog # CA10064-248
Fournisseur #:  CC-2555
PrEC – Human Prostate Epithelial Cells
Catalog # CA10064-248
CAS Number:  
PrEC – Human Prostate Epithelial Cells
PrEC – Human Prostate Epithelial Cells
PrEC – Human prostate epithelial cells, ≥500000 cells
CAS Number:  
Price
$2,283.10
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Prostate cells provide a glandular function in the body by generating fluid which serves several functions in reproduction. Lonza's Human Prostate Cells are available from epithelial, smooth muscle, and stromal tissue. Each cell type has an optimized growth media for best results in culture.
  • Guaranteed through 15 population doublings
  • Test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Prostate Epithelial Cells (PrEC) are guaranteed through 15 population doublings and stain positive for cytokeratin (clone 8.13). Cryopreserved PrEC are shipped in first or second passage.
Applications: Benign prostatic hyperplasia (BPH), Prostitis, Prostate cancer, Procreation research, Drug discovery and Male prostate physiology.
Delivery information: Content: 1 × Cryopreserved ampule PrEC containing ≥500000 cells. A Certificate of Analysis (CoA) is provided for each cell lot purchased.
Spécifications
Spécifications
  • Désignation:
    PrEC – Human prostate epithelial cells
  • Taille:
    ≥500000 cells