NHOst – Human Osteoblasts
NHOst – Human Osteoblasts
NHOst – Human osteoblasts, ≥500000 cells
Catalog # CA10064-216
Supplier #:  CC-2538
NHOst – Human Osteoblasts
Catalog # CA10064-216
CAS Number:  
NHOst – Human Osteoblasts
NHOst – Human Osteoblasts
NHOst – Human osteoblasts, ≥500000 cells
CAS Number:  
Price
$2,215.96
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Cryopreserved Clonetics® Normal Human Osteobalsts (NHOst) are guaranteed through 10 population doublings, stain positive for alkaline phosphatase and mineralization.
  • Ready-To-Use
  • No tissue dissection required
  • Increased data reproducibility
  • Characteristic morphology and staining tests confirm tissue origin and cell type
All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery information: Content: 1 × Cryopreserved ampule of NHOst containing ≥500000 cells. A Certificate of Analysis is provided for each cell lot purchased.
Specifications
Specifications
  • Désignation:
    NHOst – Human osteoblasts
  • Taille:
    ≥500000 cells