Nucleofector® Basic Kit for Primary Mammalian Epithelial Cells
In addition to various cell type-specific kits for primary epithelial cells, we also provide a cell group-specific basic Nucleofector® kit.
- Optimization can be performed within 1 experiment
- Detailed protocol guiding through the optimization procedure
- Optional result fine tuning with help of our Scientific Support Team
- Transfection efficiencies up to 83%
- Viabilities up to 98%
This kit is suited to determine the optimal transfection conditions for any mammalian smooth muscle cell type that lacks a cell type specific optimized protocol for the Nucleofector® 2b device (or older generations I or II). In a first experiment a selection of different Nucleofector® programs is tested in combination with the basic Nucleofector® solution for epithelial cells.
Applications: kit suited for fibroblasts from different mammalian species and various organs, already tested for macaque dermal fibroblasts bovine fibroblasts human colon myofibroblasts mouse lung fibroblasts etc.
Delivery information: Content: 2.25 ml Nucleofector® solution for primary mammalian epithelial cells, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mM Tris pH 8.0), 25 aluminum cuvette (100 µL), 25 single use pipettes.
- Désignation:Basic Nucleofector® kit for primary mammalian epithelial cells
- No. of Reactions:25
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



