Nucleofector® Rat Neuron Kit
Rat neuron Nucleofector® kit, 25
Supplier: LONZA PHARMA - BIOSCIENCE CA
Supplier #: VPG-1003
Nucleofector® Rat Neuron Kit
Catalog # CA10064-622
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Nucleofector® 2b system (or older generations I or II) works with cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary rat neurons.
- High transfection efficiencies for primary cells
- Cells maintain functionality post transfection
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery information: Content: 2.25 ml Nucleofector® solution for rat neuron cells, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.
Specifications
Specifications
- Désignation:Rat neuron Nucleofector® kit
- No. of Reactions:25



