Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Nucleofector® Human Mesenchymal Stem Cell Kit | Avantor

Nucleofector® Human Mesenchymal Stem Cell Kit

Human mesenchymal stem cell Nucleofector® kit, 25
Catalog # CA10064-440
Fournisseur #:  VPE-1001
Nucleofector® Human Mesenchymal Stem Cell Kit
Catalog # CA10064-440
CAS Number:  
Nucleofector® Human Mesenchymal Stem Cell Kit
Nucleofector® Human Mesenchymal Stem Cell Kit

Human mesenchymal stem cell Nucleofector® kit, 25

CAS Number:  
Price
$1,333.13
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary human mesenchymal stem cells (MSCs).

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

 : Content: 2.25 ml Nucleofector® solution for human mesenchymal cells, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.

Spécifications

Spécifications

  • Désignation:
    Human mesenchymal stem cell Nucleofector® kit
  • No. of Reactions:
    25
  • Cat. No.:
    CA10064-440

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.