Nucleofector® Mouse B Cell Kit | Avantor
Nucleofector® Mouse B Cell Kit
Mouse B cell Nucleofector® kit, 25
Catalog # CA10064-126
Fournisseur #:  VPA-1010
Nucleofector® Mouse B Cell Kit
Catalog # CA10064-126
CAS Number:  
Nucleofector® Mouse B Cell Kit
Nucleofector® Mouse B Cell Kit
Mouse B cell Nucleofector® kit, 25
CAS Number:  
Price
$1,333.13
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary mouse B cells.
  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
"Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery information: Content: 2.25 ml Nucleofector® solution for mouse B cells, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.
Spécifications
Spécifications
  • Désignation:
    Mouse B cell Nucleofector® kit
  • No. of Reactions:
    25