FitMe™ Lab Jackets
Lavender, XXL
Supplier: Dukal
Fournisseur #: UGJ-6504-XXL
Jump to:
Product Details & Documents
FitMe™ lab jackets feature a unique adjustable waistband design that provides a true custom fit.
- Knit cuffs and collar
- Snap front closure
- Premium SMS material
- 2 hip pockets, 1 chest pocket
- Washable and autoclavable up to 5 times
Available in a wide variety of colors, FitMe™ lab jackets are manufactured from premium SMS material, and are supplied non sterile.
Packaging: Lab jackets are packaged 10 per bag, 5 bags per case.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Taille de confection:XXL
- Color:Lavender
- Cat. No.:76582-128
- Sleeve or leg length:Long
- Closure type:Press studs / snaps
- Fabric:Premium SMS