Sterilization Pouches, Heat Seal Paper
HALYARD* paper-film single-use sterilization pouches are intended to be used to enclose another medical device, in a single or double pouch configuration, that is to be sterilized by a health care professional via steam and EO.
- Single-use disposable
- Premium 68 GSM paper consisting entirely of primary virgin fiber
- Excellent bacterial barrier properties that allow proper sterilant penetration
- Provide exceptional clean peeling performance and high seal strength
- Multilayer transparent film provides puncture and abrasion resistance plus a high-quality seal
- Combination of paper and film creates a high-performance, durable sterile barrier system that provides a reliable clean peel
- Paper-Film single-use pouches are printed with sterilization process indicators for steam and EO methods
: CE marked and comply with the international norms and standards ISO 11607 and EN 868.
Frequently Bought Together