AlkaGent Alkaline Cage Wash
Fournisseur: Unimed-Midwest
Certificats
À propos de cet article
Cage washing is a critical step in ensuring the safest possible biomedical environment for your staff, animals, and valuable research. Explore our portfolio of professional enzyme, alkaline, and acid cage wash detergent solutions for your life sciences facility.
- Versatile, low-foaming detergent
- Effectively removes blood and other bio-burden from surfaces
- Maintains high pH level for effective cleaning and corrosion protection of equipment
- Unique buffer/polymer system provides two modes of cleaning performance
- Rust inhibitors reduce tarnishing and corrosion
- Prevents bio-burden from depositing on cages and other equipment
- Biodegradable and phosphate-free
For use in both automated cage washers, manual use on a variety of equipment and ultrasonic units within the life science, veterinary, and research industries.
Frequently Bought Together