À propos de cet article
Produced and processed under cleanroom conditions to ensure contamination-free analysis.
- Perfect fit for Transferpette® micropipettes and suitable for other popular pipette brands
- Made from high-purity, additive-free PP
- Manufactured without lubricants and cadmium-free pigments; no parting agents or demolding aides are used
BRAND® pipette tips offer proven functionality for reliable and precise results. Tips are colorless and thin-walled, with visible graduations. Tips and TipBoxes are autoclavable at 121 °C.
Bulk tips come in convenient resealable bags. The TipBox tips are neatly packaged in a sturdy box with a hinged lid for added convenience.
The TipRacks are palletized and are designed to fit perfectly into empty TipBoxes. Sterile TipRacks include a transfer aid for contamination-free transfers into the TipBox. Racked tips come in a distinctive purple-colored tip tray for easy recognition.
All tips are manufactured under control room conditions with intensive in-process controls, including batch management and final product inspection.
BIO-CERT® liquid handling quality tips are DNA-, RNase-, DNase-, and pyrogen-free. They are also free of PCR-inhibitors and ATP. Sterile tips are sterilized with beta radiation and are supplied with a Certificate of Analysis.