Knife Switch, Small
Knife switch, screw posts, single pole, single throw
Catalog # 470353-956
Supplier: United Scientific Supplies
Fournisseur #: KSW001
Jump to:
Product Details & Documents
Tradiitonal lever switch for circuit experiments.
- Easy pivot lever
- Secure connection screws
Single-pole, single-throw knife switch with plastic base is suitable for low voltage experiments (less than 3.6 V). Includes thumbscrew connecting posts.
Spécifications
- Désignation:Knife switch, screw posts, single pole, single throw