Order Entry
Canada
ContactUsLinkComponent
United Scientific Economy Hall's Carriage
Catalog # 470354-332
undefined
United Scientific Economy Hall's Carriage
Catalog # 470354-332
Supplier Number:  HLCRG1-E

Specifications

  • Description:
    Economy Hall's carriage
  • Cat. no.:
    470354-332
  • L×W×H:
    Carriage: 5.25×2.25×1.5"
    Well: 2×2.25×0.95"

Specifications

About this item

Smoth rolling hall's carriage for velocity experiments.

  • Smooth rolling wheels
  • Well for holding masses
  • Includes hole for attaching string

This economy Hall’s carriage is perfect for demonstrations and investigating principles such as Newton’s laws, acceleration, velocity and friction. This economy Hall’s carriage has a durable plastic body with a roomy well for additional masses (not included). The hole on the front bumper allows you to easily connect a string for pulling the cart up a ramp. Both the body and wheels are made from high impact plastic.