Luer Lock Syringes
Luer Lock Syringes
Luer Lock Syringes
Certificates
Luer Lock Syringes
Luer Lock Syringes
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
General-purpose syringes available in 1, 3, 5, 10 and 20 ml barrel volumes for clinical and veterinary use.
  • EtO sterilized
  • Pyrogen-free
  • Highly transparent barrel for maximum visibility
  • Excellent lubrication performance
  • Plunger retention mechanism prevents accidental plunger withdrawal
These syringes are non-reactive and safe for use in medical procedures. The barrel and plunger are made from polypropylene, and the piston is made from polyisoprene. All syringes do not contain any DEHP, BPA, or styrene. This makes them suitable for clinical use including injecting fluid into the body and collecting fluid from the body.
Certifications: Meets ISO standards, FDA approved.
Packaging: Syringes are sterilized, individually wrapped.
Documents
Product Family Options
Product Information
  • Désignation
    Volume
    Graduation
    Tip Style
    Availability
    Price
  • Luer lock syringe
    1 ml
    0.05 ml
    Luer lock
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $34.29
    $34.29
    Log in for availability
    Case of 2400
    Case of 2400 - $806.50
    $806.50
    Box of 100
       $34.29
    Case of 2400
       $806.50
    Supplier Number: SLL101101
  • Luer lock syringe
    1 ml
    0.05 ml
    Luer lock
    Discontinued
    Supplier Number: SLL101101
  • Luer lock syringe
    3 ml
    0.01 ml
    Luer lock
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $35.61
    $35.61
    Log in for availability
    Case of 2400
    Case of 2400 - $854.64
    $854.64
    Box of 100
       $35.61
    Case of 2400
       $854.64
    Supplier Number: SLL101103
  • Luer lock syringe
    5 ml
    0.2 ml
    Luer lock
    Discontinued
    Supplier Number: -
  • Luer lock syringe
    5 ml
    0.2 ml
    Luer lock
    Discontinued
    Supplier Number: SLL101105
  • Luer lock syringe
    10 ml
    0.2 ml
    Luer lock
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $61.36
    $61.36
    Log in for availability
    Case of 1200
    Case of 1200 - $736.32
    $736.32
    Box of 100
       $61.36
    Case of 1200
       $736.32
    Supplier Number: SLL101110
  • Luer lock syringe
    20 ml
    1 ml
    Luer lock
    Discontinued
    Supplier Number: SLL101120
  • Luer lock syringe
    20 ml
    1 ml
    Luer lock
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 50
    Box of 50 - $48.68
    $48.68
    Log in for availability
    Case of 800
    Case of 800 - $763.38
    $763.38
    Box of 50
       $48.68
    Case of 800
       $763.38
    Supplier Number: SLL101120