Staticide® 7060 Polyester Swab
Medical-grade foam, 6.3", Narrow paddlehead
Catalog # 77200-552
Supplier: ACL Staticide
Fournisseur #: 7060
Jump to:
Product Details & Documents
Robust and reliable knit swab for cleanroom applications.
- Non-abrasive, double-layer polyester knit tip
- Ultra-low particle; lint-free
- Free of silicon, amides, and DOP contamination
- White polypropylene handles
- Class 10 laundered and packaged
- ISO Class 5 suitable
The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Spécifications
- Longueur:6.3"
- Matière:Medical-grade foam
- Tip Style:Narrow paddlehead