Staticide® 7255 Polyester Swab with ESD Handle
Catalog # 77200-560
Supplier: ACL Staticide
Specifications
- Length:2.75"
- Material:Polyester knit
- Cat. no.:77200-560
- Tip Style:Round
Specifications
About this item
Robust and reliable knit swab for cleanroom applications that require static control.
- Ultra-low particle; lint-free
- Free of silicon, amides, and DOP contamination
- Blue polypropylene handles have a resistance of 10⁸ to 10¹° ohms
- Class 10 laundered and packaged
- ISO Class 5 suitable
The 7255 polyester swab features a non-abrasive, double-layer polyester knit tip. The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.