Staticide® 7600 IPA Cleaning Wipes
Presaturated isopropyl alcohol wipes in a convenient pop-up canister. The 7600 is the perfect choice for all general cleaning applications where removing contaminants are of concern. Ideal for removing solder paste, ink, and adhesives.
- 70% high-purity electronics-grade isopropyl alcohol and 30% deionized water
- Supplied in a convenient pop-up container
- Removes oil, grease, ionic, and non-ionic residues
- 5×8" perforated wipes
- 54% cellulose/46% polyester hydroentangled low-lint fabric
- Made in the USA
Replacement rolls include 100 presaturated wipes.
Frequently Bought Together