Staticide® Snap Kits for ESD Mats
Fournisseur: ACL Staticide
Certificats
À propos de cet article
Create a grounding point for ESD-safe dissipative mats.
- 10 mm female socket
- 3.2 mm (0.125") center hole in socket
- Accomodates mat thicknesses up to 3 mm
- Nickel finish
- Fits the 8802 Pres-N-Snap tool
Once the snap fastener is installed on the 2-layer dissipative mat, grounding cords equipped with a 1 cm stud will clip into place.
Frequently Bought Together