Staticide® 8124 Disposable Heel Strap
Disposable heel strap, Yellow/black
Catalog # 77200-602
Supplier: ACL Staticide
Fournisseur #: 8124
Jump to:
Product Details & Documents
The easy to use ACL staticide disposable heel strap is an economical solution for plant visitors and other temporary workers who enter electrostatic protected areas.
- 99% polyester, 1% conductive yarn
- Resistance: 10⁵ ohms
These disposable heel straps are packaged 100 strips per bag.
Spécifications
- Color:Yellow/black
- Désignation:Disposable heel strap