Staticide® 8120 Economy Heel Grounder
77200-598
- Color:Black
- Description:Economy heel grounder
- Cat. no.:77200-598
The economy heel grounder is a cost-effective solution offering form-fitting comfort.
- Two-layer heel rubber
- Fits a variety of footwear
- 1-meg ohm resistor
- Black conductive exterior: 10⁴ ohms
This economy heel grounder features a conductive ground ribbon measuring 14.5×0.05", and can be secured with an adjustable hook and loop fabric with a D-ring tie-down. It is specifically designed for use by grounding employees on the move in electronic manufacturing areas.