Staticide® Universal Grounding Cord with Adapters
Catalogue # 77200-574
Fournisseur: ACL Staticide
Spécifications
- Description:Universal grounding cord with adapters
- Cat. no.:77200-574
Spécifications
À propos de cet article
All-purpose 10' ground cord built with a 1-megohm resistor.
- 10' coated high-strength tensile wire
- 10 mm female socket-to-banana cable
- Ring-style terminal lug (with banana jack)
- 10 mm stud-to-socket stack snap (with banana jack)
- 10 mm stud-to-stud stack snap (with banana jack)
- Alligator clip (with banana jack)
Supplied with four conversion adapters.