ContactUsLinkComponent
Staticide® 7022 Polyurethane Foam Swab
Catalog # 77200-550
Supplier:  ACL Staticide
undefined
Staticide® 7022 Polyurethane Foam Swab
Catalog # 77200-550
Supplier:  ACL Staticide
Supplier Number:  7022
Specifications
  • Length:
    5.04"
  • Material:
    Medical-grade foam
  • Cat. no.:
    77200-550
  • Tip Style:
    Large paddlehead
Specifications
About this item
Medical-grade foam tipped swab with superior solvent capacity.
  • High solvent capacity
  • Non-abrasive medical-grade foam
  • White polypropylene handles
  • Class 10 laundered and packaged
  • ISO Class 4-7 compatible
The swab construction features ultra sonic welding, with no adhesives or binders utilized.