Staticide® 7250 Microfiber Foam Swabs with ESD Handle
Catalogue # 77200-558
Fournisseur: ACL Staticide
Spécifications
- Length:2.75"
- Material:Medical-grade foam wrapped in microfiber knit
- Cat. no.:77200-558
- Tip Style:Small round
Spécifications
À propos de cet article
ESD-safe handle and and microfiber head for exceptionally clean critical cleaning.
- Ultra-low particles, lint-free
- Non-abrasive
- ESD rigid handle, measures 10e8 -10e10 ohms
- Class 10 cleanroom laundered
- ISO Class 4-7 compatible
The 7250 microfiber foam swab features a small round tip constructed of medical-grade foam wrapped in microfiber knit. The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.