Staticide® ACL Microfiber Wipes | Avantor
Staticide® ACL Microfiber Wipes
Supplier:   ACL Staticide
Staticide® ACL Microfiber Wipes
Supplier:  ACL Staticide
Certificates
Staticide® ACL Microfiber Wipes
Staticide® ACL Microfiber Wipes
2 Options Available Below
Product Details & Documents
The ACL Staticide microfiber wipe for critical task cleaning is made from hydro-entangled split microfiber. This material provides excellent pick-up and removal of contaminants, whether used wet or dry.
  • 80% polyester/20% nylon hydro-entangled blend
  • Binder-free, no chemical residue
  • Ultra-low particle and fiber release
  • Knife-cut edge
Offering excellent durability, strength, and liquid absorbancy, the Microfiber wipes are suitable for use in ISO Class 5 (Class 100) environments.
Product Family Options
Informations sur le produit
  • Matière
    Packaging
    W×L
    Availability
    Price
  • 80% polyester/20% nylon hydroentangled microfiber blend
    600 wipes/bag, 5 bags/case
    10.2×10.2 cm (4×4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 600
    Case of 600 - $556.27
    $556.27
    Case of 600
       $556.27
    Supplier Number: 8404MF
  • 80% polyester/20% nylon hydroentangled microfiber blend
    150 wipes/bag, 6 bags/case
    22.9×22.9 cm (9×9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 150
    Case of 150 - $502.26
    $502.26
    Case of 150
       $502.26
    La quantité minimale de commande est de 1
    Supplier Number: 8409MF