Staticide® 8122 Premium Heel Grounder
Catalog # 77200-600
Supplier: ACL Staticide
Specifications
- Color:Black
- Description:Premium heel grounder
- Cat. no.:77200-600
Specifications
About this item
The premium heel grounder provides added conductivity, strength and longevity thanks to an extra-wide grounding strap.
- Two-layer heel rubber
- Wide cup-style
- Reinforced with steel rivets
- 1-meg ohm resistor
- Black conductive exterior: 10⁴ ohms
This premium heel grounder features a conductive ground ribbon measuring 14.5×0.05", and can be secured with an adjustable hook and loop fabric. It is specifically designed for use by grounding employees on the move in electronic manufacturing areas.