Staticide® Common Point Ground Cord for ESD Mats, 10 mm Stud with Dual Port
Catalog # 77200-570
Supplier: ACL Staticide
Specifications
- Description:Common point ground cord for ESD mats
- Cat. no.:77200-570
Specifications
About this item
The 8091 common point mat ground cord snaps onto a groundable point on an ESD-safe mat.
- 10' coated high-strength tensile wire
- 1-meg ohm resistor
- 10mm male stud connection to mats
- Ring-style termination for connecting to ground point
- Equipped with two banana jack plug-ins
This ground point features two plug-in banana jacks for wrist straps while connecting mats to a ground source.