Staticide® Two-Point Resistance Checker
Two-point resistance checker, 2.8×5.1×1.4"
Catalog # 77200-718
Supplier: ACL Staticide
Supplier #: ACL306
Jump to:
Product Details & Documents
The Staticide ACL 306 is a convenient 2-point probe resistance tester for measuring small areas.
- Works independent of a megohmmeter
- Measuring range: 10³ to 10¹² ohms
- Measures 0.375" wide
- Made in USA
Delivery information: Includes calibration certificate.
Specifications
- Désignation:Two-point resistance checker
- W×D×H:2.8×5.1×1.4"