Staticide® Checker Verification Board
Checker verification board, accessory standard set, 5×3.25×0.19"
Catalog # 77200-720
Supplier: ACL Staticide
: ACL307
Jump to:
Product Details & Documents
The ACL 307 checker board is a convenient verification unit for ensuring the ACL 306 two-point resistance tester is accurate.
- Metal pads with 1% resistor
- Measuring range: 10³ to 10¹° ohms
- Measurements over 10¹° ohms will indicate ≥10¹¹
- Made in USA
: Includes calibration certificate.
Spécifications
- Désignation:Checker verification board, accessory standard set
- W×D×H:5×3.25×0.19"