Staticide® Pres-N-Snap Installation Hand Tool
Pres-N-Snap Tool for grounding ESD mats, Cast aluminum
Catalog # 77200-608
Supplier: ACL Staticide
: 8802
Jump to:
Product Details & Documents
Heavy-duty clamping tool for attaching grounding point snaps on ESD-safe dissipative mats.
- Heavy-duty cast aluminum construction
- Made in USA
Includes attaching dies for 1 cm snaps, and ten pairs of female sockets and eyelets.
Spécifications
- Désignation:Pres-N-Snap Tool for grounding ESD mats
- Matière:Cast aluminum