Eisco Evaporating Flasks
 :   Wards
Eisco Evaporating Flasks
 :  Wards
Eisco Evaporating Flasks
Eisco Evaporating Flasks
Evaporating flasks with ground sockets made of borosilicate 3.3 glass.
  • Withstands high temperatures and thermal shocks
  • Ungraduated, with round bottom
  • Features ground socket
  • Used with rotary evaporators and vacuum applications that vaporize solvents
Evaporating flasks for use with rotary evaporators and vacuum applications that vaporize solvents. Made of durable, high quality, heavy duty 3.3 borosilicate glass with the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Product Family Options
  • Capacité
    ST Joint
  • 100 ml
    24/29
    Each
    Each - $18.95
    $18.95
    Each
       $18.95
     : CH0435A
  • 250 ml
    24/29
    Each
    Each - $27.95
    $27.95
    Each
       $27.95
     : CH0435B
  • 250 ml
    29/32
    Each
    Each - $30.70
    $30.70
    Each
       $30.70
     : CH0435C
  • 500 ml
    24/29
    Each
    Each - $37.30
    $37.30
    Each
       $37.30
     : CH0435D
  • 500 ml
    29/32
    Each
    Each - $37.90
    $37.90
    Each
       $37.90
     : CH0435E
  • 1000 ml
    24/29
    Each
    Each - $40.45
    $40.45
    Each
       $40.45
     : CH0435F
  • 1000 ml
    29/32
    Each
    Each - $47.40
    $47.40
    Each
       $47.40
     : CH0435G