Borosil® Erlenmeyer Flasks, Narrow Neck, Graduated
À propos de cet article
These narrow mouthed flasks are made of clear borosilicate 3.3 glass.
- Uniform wall thickness
- Beaded rim
- Graduated
- Type 1 glass
Borosil® flasks are designed to deliver a combination of thermal shock resistance along with physical strength and resistance to chemical attack. These flasks comply with IS 1381 (Part I), DIN/ ISO 1773 and are manufactured to withstand the demands of the modern laboratory.
The uniform wall thickness distribution makes these flasks ideal for heating. The conical form makes these flasks also suitable for mixing liquids. The easy-to-read scale and large labeling field helps to ensure for easy marking. 25 ml to 5 L are graduated in a ceramic white color.
: ISO 1773
11 Options Available Below
Product Details & Documents
These narrow mouthed flasks are made of clear borosilicate 3.3 glass.
- Uniform wall thickness
- Beaded rim
- Graduated
- Type 1 glass
Borosil® flasks are designed to deliver a combination of thermal shock resistance along with physical strength and resistance to chemical attack. These flasks comply with IS 1381 (Part I), DIN/ ISO 1773 and are manufactured to withstand the demands of the modern laboratory.
The uniform wall thickness distribution makes these flasks ideal for heating. The conical form makes these flasks also suitable for mixing liquids. The easy-to-read scale and large labeling field helps to ensure for easy marking. 25 ml to 5 L are graduated in a ceramic white color.
: ISO 1773
Product Family Options
Informations sur le produit
- CapacitéIntervalle de graduationPlage de graduationHauteurØ ext.Neck I.D.AvailabilityPrice
- 10 ml——60 mm37 mm15 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 30Case of 30 - $280.10$280.10Prix promotionnel pour les nouveaux clientsCase of 30$280.10Supplier Number: 4980006
- 25 ml——75 mm40 mm22 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 40Case of 40 - $367.92$367.92Prix promotionnel pour les nouveaux clientsCase of 40$367.92Supplier Number: 4980009
- 50 ml10 ml10 - 50 ml90 mm51 mm22 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $959.22$959.22Prix promotionnel pour les nouveaux clientsCase of 100$959.22Supplier Number: 4980012
- 100 ml50 ml40 - 100 ml110 mm66 mm28 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $939.68$939.68Prix promotionnel pour les nouveaux clientsCase of 100$939.68Supplier Number: 4980016
- 150 ml50 ml50 - 100 ml124 mm72 mm28 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $1,246.90$1,246.90Prix promotionnel pour les nouveaux clientsCase of 100$1,246.90Supplier Number: 4980018
- 250 ml100 ml200 - 500 ml145 mm85 mm34 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 40Case of 40 - $412.83$412.83Prix promotionnel pour les nouveaux clientsCase of 40$412.83Supplier Number: 4980021
- 500 ml50 ml100 - 250 ml180 mm105 mm38 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 40Case of 40 - $510.72$510.72Prix promotionnel pour les nouveaux clientsCase of 40$510.72Supplier Number: 4980024
- 1000 ml (1 L)200 ml400 - 1000 ml220 mm131 mm42 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 20Case of 20 - $398.76$398.76Prix promotionnel pour les nouveaux clientsCase of 20$398.76Supplier Number: 4980029
- 2000 ml (2 L)400 ml800 - 2000 ml280 mm166 mm52 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 20Case of 20 - $805.89$805.89Prix promotionnel pour les nouveaux clientsCase of 20$805.89Supplier Number: 4980030
- 3000 ml (3 L)500 ml1000 - 3000 ml310 mm187 mm48 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 4Case of 4 - $493.85$493.85Prix promotionnel pour les nouveaux clientsCase of 4$493.85Supplier Number: 4980031
- 5000 ml (5 L)500 ml1000 - 5000 ml365 mm220 mm50 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 2Case of 2 - $387.22$387.22Prix promotionnel pour les nouveaux clientsCase of 2$387.22Supplier Number: 4980033