Eisco Plastic Separatory Funnels
 :   Wards
Eisco Plastic Separatory Funnels
 :  Wards
Eisco Plastic Separatory Funnels
Eisco Plastic Separatory Funnels
Polypropylene separatory funnels have a screw cap and key stopcock fitted with a self-lubricating PTFE plug.
  • Made of premium-quality polypropylene plastic, with superior optical clarity, chemical durability, and heat tolerance
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Polypropylene key stopcock fitted with a self-lubricating PTFE plug push-fitted to the stem of the funnel for easy and precise flow control during separation procedures
  • Leak-proof threaded screw cap for protecting contents
  • Ungraduated
Separatory funnels are made of high-quality polypropylene plastic, with excellent clarity and strength. Can withstand temperatures up to 135°C. Autoclave, microwave and dishwasher safe. Excellent resistance to strong acids, alcohols and bases. It includes a leak-proof, threaded screw cap and key stopcock, fitted with a self-lubricating PTFE plug that is pushed fitted to funnel stem. Ungraduated.
Product Family Options
  • Capacité
  • 100 ml
    Each
    Each - $28.40
    $28.40
    Each
       $28.40
     : CH0480A
  • 250 ml
    Each
    Each - $33.20
    $33.20
    Each
       $33.20
     : CH0480B
  • 500 ml
    Each
    Each - $39.45
    $39.45
    Each
       $39.45
     : CH0480C