ContactUsLinkComponent
VWR® Tissue Flotation Water Bath, 230 V
  80086-984
Product available on GSA Advantage®
Irregular Voltage
undefined
VWR® Tissue Flotation Water Bath, 230 V
  80086-984
Product available on GSA Advantage®
Irregular Voltage
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Temperature range:
    35 to 65°C (95 to 149°F)
  • Description:
    Tissue Flotation Bath, 230V, 50/60Hz, 190W
  • Electrical:
    230V, 50/60Hz, 190W
  • Temperature stability:
    ±1°C
  • Ext. W×D×H:
    28 dia. x 9.2H cm (11 x 35/8"), Rim: 3.2 cm (11/4")
  • Int. W×D×H:
    Chamber: 21 dia. x 5D cm (81/4 x 2")
  • Cat. no.:
    80086-984
Typical use of tissue flotation water bath is in the preparation of tissue paraffin sections and the elimination of wrinkles and distortions of the tissue sections.
  • 230 V model, not for use in USA
  • Round and seamless
  • Sheet aluminum construction
  • Bath surface is black ceramic-coated steel
  • Improved visibility of tissue sections
Designed especially for tests and procedures using tissue cultures.
Round, seamless, sheet aluminum bath is insulated with fiberglass. It features a baked gray enamel exterior and a cover. Bath surface is black ceramic-coated steel, which offers improved visibility of tissue sections. The warm broad edge of the bath is a convenient drying surface for processed slides. Complete with thermostat reference dial and knob, thermometer holder, pilot light, 18-gauge three-wire rubber safety cord and plug, and skid-resistant feet.
 : 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (80086-982) is also available, please see separate product page.