Eisco Plastic Filter Funnels
Eisco Plastic Filter Funnels
Supplier:   Wards
Eisco Plastic Filter Funnels
Supplier:  Wards
Certificates
Eisco Plastic Filter Funnels
Eisco Plastic Filter Funnels
2 Options Available Below
Product Details & Documents
Chemically-resistant filter funnels made of high quality polypropylene plastic.
  • Excellent resistance against acids, alcohols, aldehydes, bases, and hydrocarbons
  • Useful for separating solids from liquids, as well as for easier transfers of liquids to narrow mouthed containers
  • Autoclavable and microwavable
Packaging: Pack of 1 or 10.
Product Family Options
Informations sur le produit
  • Taille
    Ø
    Stem Length
    Availability
    Price
  • 10
    50 mm
    50 mm
    Discontinued
    Supplier Number: -
  • 10
    62 mm
    62 mm
    Discontinued
    Supplier Number: -
  • 10
    75 mm
    76 mm
    Discontinued
    Supplier Number: -
  • 10
    100 mm
    80 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $20.65
    $20.65
    Each
       $20.65
    Supplier Number: CH0454D10
  • 1
    150 mm
    110 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6.70
    $6.70
    Each
       $6.70
    Supplier Number: CH0454E