Single Frosted Microscope Slides | Avantor
Single Frosted Microscope Slides
Single Frosted Microscope Slides
Certificates
Single Frosted Microscope Slides
Single Frosted Microscope Slides
1 Options Available Below
Product Details & Documents
Four E's frosted microscope slides made from high quality soda glass or super white glass.
  • Pre-cleaned, ready to use
  • Frosted single end, single side
  • Grounded edges, 45° corners
  • Autoclavable
The 20 mm wide marking surface can be marked by the label method, 2B pencil, and marker. The marking will not be easily wiped or washed off, and is suitable for use with common chemical reagents and dyes in laboratories. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.
Product Family Options
Informations sur le produit
  • Matière
    Épaisseur
    L×l
    Availability
    Price
  • Soda glass
    1.0 mm
    75×25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 72
    Box of 72 - $11.80
    $11.80
    Log in pour la disponibilité
    Case of 3600
    Case of 3600 - $588.91
    $588.91
    Box of 72
       $11.80
    Case of 3600
       $588.91
    Supplier Number: SLD145210
  • Super white glass
    1.0 mm
    75×25 mm
    Discontinued
    Supplier Number: SLD245210