Staticide® ESD Knit Gloves
Staticide® ESD Knit Gloves
Supplier:   ACL Staticide
Staticide® ESD Knit Gloves
Supplier:  ACL Staticide
Certificates
Staticide® ESD Knit Gloves
5 Options Available Below
Product Details & Documents
These ESD Knit gloves are ideal for inspection and assembly.
  • Seamless knit construction
  • Nylon conductive carbon fiber blend yarn
  • Low moisture absorption
  • GLK CAFE RGP: 10⁷ ohms
These micro-knit static control gloves offer superb comfort and durability. Ideal for inspection and assembly work. Ideal for inspection and assembly work when static control is required.
 : Ordered by case, six packs (36 pairs of gloves) per case.
 : Each pack contains six pairs.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 77200-752
  • 77200-750
  • 77200-748
  • 77200-764
  • 77200-746
  • Ambidextrous/Hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Taille du gant
    Glove Color
    Longueur
    Épaisseur
    Availability
    Price
  • Staticide ESD knit gloves with white rib
    S
    White
    25.4 cm (10")
    13 gauge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-S
    Spécifications:
    More
    Specifications
    Cat. No.
    77200-752
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • Staticide ESD knit gloves with green rib
    M
    White
    25.4 cm (10")
    13 gauge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-M
    Spécifications:
    More
    Specifications
    Cat. No.
    77200-750
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • Staticide ESD knit gloves with brown rib
    L
    White
    25.4 cm (10")
    13 gauge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-L
    Spécifications:
    More
    Specifications
    Cat. No.
    77200-748
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • Staticide ESD knit gloves with yellow rib
    XL
    White
    25.4 cm (10")
    13 gauge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-XL
    Spécifications:
    More
    Specifications
    Cat. No.
    77200-764
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • Staticide ESD knit gloves with gray rib
    XXL
    White
    25.4 cm (10")
    13 gauge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $216.75
    $216.75
    Case of 500
       $216.75
    Supplier Number: GLK-2XL
    Spécifications:
    More
    Specifications
    Cat. No.
    77200-746
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile