Screen Sieve Set 6
Screen Sieve Set 6
Supplier:   Avantor
Screen Sieve Set 6
Supplier:  Avantor
Certificates
Screen Sieve Set 6
Screen Sieve Set 6
7 Options Available Below
Product Details & Documents
An economical set of durable sieve that will hold up to tough use.
  • Set includes 6 popular mesh sizes
  • The frame is made from durable steel
  • A top and bottom cover are provided for storage
  • Each screan is labeled with the mesh size
  • 15 cm diameter for all sieves
The process of screening allows students to study the make up of the materials they have collected. The screening process athat aid in the recognition of the different parts of a specimen. The screen sieves are a valuable tool used in this process. This set features steel screens with six different sizes of mess that include: 5, 10, 35, 60, 120, 230. A top and bottom is also provided to enclose the set.
Ordering information: Each screen can be purchased individual.
Product Family Options
Product Information
  • Désignation
    Mesh Size
    Availability
    Price
  • Screen sieve
    5
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: -
  • Screen sieve set of 6
    Log in for availability
    Log in for availability
    Each
    Each - $137.46
    $137.46
    Each
       $137.46
    Supplier Number: SS-15-6
  • Screen sieve
    10
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: SSC-15-10
  • Screen sieve
    35
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: -
  • Screen sieve
    60
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: -
  • Screen sieve
    120
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: -
  • Screen sieve
    230
    Log in for availability
    Log in for availability
    Each
    Each - $22.91
    $22.91
    Each
       $22.91
    Supplier Number: SSC-15-230